6-Chloro-N-cyclohexyl-N-methyl-4-pyrimidinamine

Supplier: Matrix Scientific

051848-500MG 051848-2.5G
101897-284EA 364.62 USD
101897-284 101897-286
6-Chloro-N-cyclohexyl-N-methyl-4-pyrimidinamine
6-Chloro-N-cyclohexyl-N-methyl-4-pyrimidinamine

Hazard: Irritant

Formula: C₁₁H₁₆ClN₃
MDL Number: MFCD13561824
CAS Number: 1219961-12-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR