4-Chloro-6-(4-methyl-1-piperazinyl)pyrimidine

Supplier: Matrix Scientific

051842-500MG 051842-2.5G
101897-260EA 192.31 USD
101897-260 101897-262
4-Chloro-6-(4-methyl-1-piperazinyl)pyrimidine
4-Chloro-6-(4-methyl-1-piperazinyl)pyrimidine

Hazard: Irritant

Formula: C₉H₁₃ClN₄
MDL Number: MFCD05739691
CAS Number: 27174-60-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR