4-Chloro-6-(4-methyl-1-piperidinyl)pyrimidine

Supplier: Matrix Scientific

051841-500MG 051841-2.5G
101897-256EA 364.62 USD
101897-256 101897-258
4-Chloro-6-(4-methyl-1-piperidinyl)pyrimidine
4-Chloro-6-(4-methyl-1-piperidinyl)pyrimidine

Hazard: Irritant

Formula: C₁₀H₁₄ClN₃
MDL Number: MFCD13561821
CAS Number: 1219967-85-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR