4-Chloro-6-(3-methyl-1-piperidinyl)pyrimidine
Supplier: Matrix Scientific
051840-500MG
051840-2.5G
101897-252EA
364.62
USD
101897-252
101897-254
4-Chloro-6-(3-methyl-1-piperidinyl)pyrimidine
4-Chloro-6-(3-methyl-1-piperidinyl)pyrimidine
Hazard: Irritant
Formula:
C₁₀H₁₄ClN₃ |
MDL Number:
MFCD13561820 CAS Number: 1220036-25-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...