4-Chloro-6-(2-methyl-1-piperidinyl)pyrimidine

Supplier: Matrix Scientific

051839-500MG 051839-2.5G
101897-248EA 364.62 USD
101897-248 101897-250
4-Chloro-6-(2-methyl-1-piperidinyl)pyrimidine
4-Chloro-6-(2-methyl-1-piperidinyl)pyrimidine

Hazard: Irritant

Formula: C₁₀H₁₄ClN₃
MDL Number: MFCD13561819
CAS Number: 1219961-07-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR