N-Butyl-6-chloro-N-methyl-4-pyrimidinamine

Supplier: Matrix Scientific

051836-500MG 051836-2.5G
101897-236EA 364.62 USD
101897-236 101897-238
N-Butyl-6-chloro-N-methyl-4-pyrimidinamine
N-Butyl-6-chloro-N-methyl-4-pyrimidinamine

Hazard: Irritant

Formula: C₉H₁₄ClN₃
MDL Number: MFCD13561816
CAS Number: 1219967-79-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR