1-(6-Chloro-4-pyrimidinyl)-3-pyrrolidinol

Supplier: Matrix Scientific

051835-500MG 051835-2.5G
101897-232EA 364.62 USD
101897-232 101897-234
1-(6-Chloro-4-pyrimidinyl)-3-pyrrolidinol
1-(6-Chloro-4-pyrimidinyl)-3-pyrrolidinol

Hazard: Irritant

Formula: C₈H₁₀ClN₃O
MDL Number: MFCD09879092
CAS Number: 959240-48-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR