4-(6-Chloro-4-pyrimidinyl)morpholine

Supplier: Matrix Scientific

Synonyms: 4-(6-Chloropyrimidin-4-yl)morpholine

051834-500MG 051834-2.5G
101897-228EA 364.62 USD
101897-228 101897-230
4-(6-Chloro-4-pyrimidinyl)morpholine
4-(6-Chloro-4-pyrimidinyl)morpholine

Hazard: Irritant

Formula: C₈H₁₀ClN₃O
MW: 199.64 g/mol
MDL Number: MFCD00466219
CAS Number: 22177-92-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR