4-Chloro-6-(1-piperidinyl)pyrimidine

Supplier: Matrix Scientific

051833-500MG 051833-2.5G
101897-224EA 364.62 USD
101897-224 101897-226
4-Chloro-6-(1-piperidinyl)pyrimidine
4-Chloro-6-(1-piperidinyl)pyrimidine

Hazard: Irritant

Formula: C₉H₁₂ClN₃
MDL Number: MFCD00834993
CAS Number: 1722-14-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR