N-(6-Chloro-4-pyrimidinyl)-N,N-diethylamine

Supplier: Matrix Scientific

Synonyms: 4-Chloro-6-(diethylamino)pyrimidine

051831-500MG 051831-2.5G
101897-216EA 364.62 USD
101897-216 101897-218
N-(6-Chloro-4-pyrimidinyl)-N,N-diethylamine
N-(6-Chloro-4-pyrimidinyl)-N,N-diethylamine

Hazard: Irritant

Formula: C₈H₁₂ClN₃
MW: 185.66 g/mol
MDL Number: MFCD09800978
CAS Number: 951885-37-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR