6-Chloro-N,N-dimethylpyrimidin-4-amine

Supplier: Matrix Scientific

Synonyms: (6-Chloro-pyrimidin-4-yl)-dimethyl-amine

051829-500MG 051829-2.5G
101897-208EA 364.62 USD
101897-208 101897-210
6-Chloro-N,N-dimethylpyrimidin-4-amine
6-Chloro-N,N-dimethylpyrimidin-4-amine

Hazard: Irritant

Formula: C₆H₈ClN₃
MW: 157.6000 g/mol
MDL Number: MFCD09756954
CAS Number: 31058-83-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR