N2-Benzyl-N2-ethyl-2,5-pyridinediamine

Supplier: Matrix Scientific

051827-500MG 051827-2.5G
101897-200EA 364.62 USD
101897-200 101897-202
N2-Benzyl-N2-ethyl-2,5-pyridinediamine
N2-Benzyl-N2-ethyl-2,5-pyridinediamine

Hazard: Irritant

Formula: C₁₄H₁₇N₃
MDL Number: MFCD09882492
CAS Number: 1017143-43-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR