2-[4-(5-Amino-2-pyridinyl)-1-piperazinyl]-1-ethanol

Supplier: Matrix Scientific

Synonyms: 2-(4-(5-Aminopyridin-2-yl)piperazin-1-yl)ethanol

051823-500MG 051823-2.5G
101897-184EA 364.62 USD
101897-184 101897-186
2-[4-(5-Amino-2-pyridinyl)-1-piperazinyl]-1-ethanol
2-[4-(5-Amino-2-pyridinyl)-1-piperazinyl]-1-ethanol

Hazard: Irritant

Formula: C₁₁H₁₈N₄O
MW: 222.29 g/mol
MDL Number: MFCD09882493
CAS Number: 1017221-34-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR