N2-Cyclohexyl-N2-ethyl-2,5-pyridinediamine

Supplier: Matrix Scientific

051822-500MG 051822-2.5G
101897-180EA 364.62 USD
101897-180 101897-182
N2-Cyclohexyl-N2-ethyl-2,5-pyridinediamine
N2-Cyclohexyl-N2-ethyl-2,5-pyridinediamine

Hazard: Irritant

Formula: C₁₃H₂₁N₃
MDL Number: MFCD09731273
CAS Number: 953904-28-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR