N2-Benzyl-N2-methyl-2,5-pyridinediamine

Supplier: Matrix Scientific

051821-500MG 051821-2.5G
101897-176EA 364.62 USD
101897-176 101897-178
N2-Benzyl-N2-methyl-2,5-pyridinediamine
N2-Benzyl-N2-methyl-2,5-pyridinediamine

Hazard: Irritant

Formula: C₁₃H₁₅N₃
MDL Number: MFCD09812641
CAS Number: 181114-67-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR