6-(2-Ethyl-1-piperidinyl)-3-pyridinamine

Supplier: Matrix Scientific

051820-500MG 051820-2.5G
101897-172EA 364.62 USD
101897-172 101897-174
6-(2-Ethyl-1-piperidinyl)-3-pyridinamine
6-(2-Ethyl-1-piperidinyl)-3-pyridinamine

Hazard: Irritant

Formula: C₁₂H₁₉N₃
MDL Number: MFCD09882495
CAS Number: 1017358-22-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR