N2-Cyclohexyl-N2-methyl-2,5-pyridinediamine

Supplier: Matrix Scientific

051819-500MG 051819-2.5G
101897-168EA 364.62 USD
101897-168 101897-170
N2-Cyclohexyl-N2-methyl-2,5-pyridinediamine
N2-Cyclohexyl-N2-methyl-2,5-pyridinediamine

Hazard: Irritant

Formula: C₁₂H₁₉N₃
MDL Number: MFCD09739184
CAS Number: 224187-13-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR