N2,N2-Dipropyl-2,5-pyridinediamine

Supplier: Matrix Scientific

051818-500MG 051818-2.5G
101897-164EA 384.62 USD
101897-164 101897-166
N2,N2-Dipropyl-2,5-pyridinediamine
N2,N2-Dipropyl-2,5-pyridinediamine

Hazard: Irritant

Formula: C₁₁H₁₉N₃
MDL Number: MFCD09044940
CAS Number: 52025-51-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR