1-(5-Amino-2-pyridinyl)-4-piperidinol

Supplier: Matrix Scientific

Synonyms: 1-(5-Aminopyridin-2-yl)piperidin-4-ol

051817-500MG 051817-2.5G
101897-160EA 230.77 USD
101897-160 101897-162
1-(5-Amino-2-pyridinyl)-4-piperidinol
1-(5-Amino-2-pyridinyl)-4-piperidinol

Hazard: Irritant

Formula: C₁₀H₁₅N₃O
MW: 193.25 g/mol
MDL Number: MFCD09728697
CAS Number: 476342-37-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR