6-(1-Azepanyl)-3-pyridinamine

Supplier: Matrix Scientific

051815-500MG 051815-2.5G
101897-152EA 364.62 USD
101897-152 101897-154
6-(1-Azepanyl)-3-pyridinamine
6-(1-Azepanyl)-3-pyridinamine

Hazard: Irritant

Formula: C₁₁H₁₇N₃
MDL Number: MFCD08700222
CAS Number: 850040-18-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR