6-(4-Benzyl-1-piperazinyl)-3-pyridinylamine

Supplier: Matrix Scientific

051814-500MG 051814-2.5G
101897-148EA 364.62 USD
101897-148 101897-150
6-(4-Benzyl-1-piperazinyl)-3-pyridinylamine
6-(4-Benzyl-1-piperazinyl)-3-pyridinylamine

Hazard: Irritant

Formula: C₁₆H₂₀N₄
MDL Number: MFCD08165811
CAS Number: 381706-46-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR