6-(4-Methyl-1-piperidinyl)-3-pyridinylamine

Supplier: Matrix Scientific

051813-500MG 051813-2.5G
101897-144EA 364.62 USD
101897-144 101897-146
6-(4-Methyl-1-piperidinyl)-3-pyridinylamine
6-(4-Methyl-1-piperidinyl)-3-pyridinylamine

Hazard: Irritant

Formula: C₁₁H₁₇N₃
MDL Number: MFCD08700205
CAS Number: 767583-34-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR