6-(3-Methyl-1-piperidinyl)-3-pyridinylamine

Supplier: Matrix Scientific

051812-500MG 051812-2.5G
101897-140EA 364.62 USD
101897-140 101897-142
6-(3-Methyl-1-piperidinyl)-3-pyridinylamine
6-(3-Methyl-1-piperidinyl)-3-pyridinylamine

Hazard: Irritant

Formula: C₁₁H₁₇N₃
MW: 191.28 g/mol
MDL Number: MFCD08700206
CAS Number: 777011-36-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR