6-(2-Methyl-1-piperidinyl)-3-pyridinylamine

Supplier: Matrix Scientific

051811-500MG 051811-2.5G
101897-136EA 364.62 USD
101897-136 101897-138
6-(2-Methyl-1-piperidinyl)-3-pyridinylamine
6-(2-Methyl-1-piperidinyl)-3-pyridinylamine

Hazard: Irritant

Formula: C₁₁H₁₇N₃
MDL Number: MFCD09050561
CAS Number: 926194-75-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR