N2,N2-Diallyl-2,5-pyridinediamine

Supplier: Matrix Scientific

051810-500MG 051810-2.5G
101897-132EA 364.62 USD
101897-132 101897-134
N2,N2-Diallyl-2,5-pyridinediamine
N2,N2-Diallyl-2,5-pyridinediamine

Hazard: Irritant

Formula: C₁₁H₁₅N₃
MDL Number: MFCD09882490
CAS Number: 1017171-94-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR