2-[(5-Amino-2-pyridinyl)(ethyl)amino]-1-ethanol

Supplier: Matrix Scientific

051809-500MG 051809-2.5G
101897-128EA 364.62 USD
101897-128 101897-130
2-[(5-Amino-2-pyridinyl)(ethyl)amino]-1-ethanol
2-[(5-Amino-2-pyridinyl)(ethyl)amino]-1-ethanol

Hazard: Irritant

Formula: C₉H₁₅N₃O
MDL Number: MFCD09882489
CAS Number: 1017221-30-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR