N2-Butyl-N2-methyl-2,5-pyridinediamine

Supplier: Matrix Scientific

051808-500MG 051808-2.5G
101897-124EA 384.62 USD
101897-124 101897-126
N2-Butyl-N2-methyl-2,5-pyridinediamine
N2-Butyl-N2-methyl-2,5-pyridinediamine

Hazard: Irritant

Formula: C₁₀H₁₇N₃
MDL Number: MFCD09808823
CAS Number: 52025-58-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR