6-(1-Piperidinyl)-3-pyridinamine

Supplier: Matrix Scientific

051806-500MG 051806-2.5G
101897-116EA 364.62 USD
101897-116 101897-118
6-(1-Piperidinyl)-3-pyridinamine
6-(1-Piperidinyl)-3-pyridinamine

Hazard: Irritant

Formula: C₁₀H₁₅N₃
MW: 177.25 g/mol
MDL Number: MFCD06447399
CAS Number: 55403-29-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR