2-[(5-Amino-2-pyridinyl)(methyl)amino]-1-ethanol

Supplier: Matrix Scientific

051805-500MG 051805-2.5G
101897-112EA 384.62 USD
101897-112 101897-114
2-[(5-Amino-2-pyridinyl)(methyl)amino]-1-ethanol
2-[(5-Amino-2-pyridinyl)(methyl)amino]-1-ethanol

Hazard: Irritant

Formula: C₈H₁₃N₃O
MDL Number: MFCD08692354
CAS Number: 4928-46-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR