N2,N2-Diethyl-2,5-pyridinediamine

Supplier: Matrix Scientific

051804-500MG 051804-2.5G
101897-108EA 364.62 USD
101897-108 101897-110
N2,N2-Diethyl-2,5-pyridinediamine
N2,N2-Diethyl-2,5-pyridinediamine

Hazard: Irritant

Formula: C₉H₁₅N₃
MDL Number: MFCD09046037
CAS Number: 34392-84-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR