6-(1-Pyrrolidinyl)-3-pyridinamine

Supplier: Matrix Scientific

Synonyms: 5-Amino-2-(pyrrolidin-1-yl)pyridine

051803-500MG 051803-2.5G
101897-104EA 150.77 USD
101897-104 101897-106
6-(1-Pyrrolidinyl)-3-pyridinamine
6-(1-Pyrrolidinyl)-3-pyridinamine

Hazard: Irritant

Formula: C₉H₁₃N₃
MW: 163.22 g/mol
MDL Number: MFCD03425745
CAS Number: 92808-19-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR