N2,N2-Dimethyl-2,5-pyridinediamine

Supplier: Matrix Scientific

Synonyms: N2,N2-Dimethylpyridine-2,5-diamine

051802-500MG 051802-2.5G
101897-100EA 384.62 USD
101897-100 101897-102
N2,N2-Dimethyl-2,5-pyridinediamine
N2,N2-Dimethyl-2,5-pyridinediamine

Hazard: Irritant

Formula: C₇H₁₁N₃
MW: 137.19 g/mol
MDL Number: MFCD08691371
CAS Number: 4928-43-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR