N-(5-Bromo-4-methyl-2-pyridinyl)-N-methyl-N-(tetrahydro-2H-pyran-4-ylmethyl)amine

Supplier: Matrix Scientific

051801-500MG 051801-2.5G
101897-096EA 364.62 USD
101897-096 101897-098
N-(5-Bromo-4-methyl-2-pyridinyl)-N-methyl-N-(tetrahydro-2H-pyran-4-ylmethyl)amine
N-(5-Bromo-4-methyl-2-pyridinyl)-N-methyl-N-(tetrahydro-2H-pyran-4-ylmethyl)amine

Hazard: Irritant

Formula: C₁₃H₁₉BrN₂O
MDL Number: MFCD13561813
CAS Number: 1220036-21-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR