1-(5-Bromo-4-methyl-2-pyridinyl)-1,2,3,4-tetrahydroquinoline
Supplier: Matrix Scientific
051799-500MG
051799-2.5G
101897-088EA
364.62
USD
101897-088
101897-090
1-(5-Bromo-4-methyl-2-pyridinyl)-1,2,3,4-tetrahydroquinoline
1-(5-Bromo-4-methyl-2-pyridinyl)-1,2,3,4-tetrahydroquinoline
Hazard: Irritant
Formula:
C₁₅H₁₅BrN₂ |
MDL Number:
MFCD13561811 CAS Number: 1219964-63-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...