1-(5-Bromo-4-methyl-2-pyridinyl)indoline
Supplier: Matrix Scientific
051798-500MG
051798-2.5G
101897-084EA
364.62
USD
101897-084
101897-086
1-(5-Bromo-4-methyl-2-pyridinyl)indoline
1-(5-Bromo-4-methyl-2-pyridinyl)indoline
Hazard: Irritant
Formula:
C₁₄H₁₃BrN₂ |
MDL Number:
MFCD13561810 CAS Number: 1219967-52-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...