2-[4-(5-Bromo-4-methyl-2-pyridinyl)-1-piperazinyl]-1-ethanol

Supplier: Matrix Scientific

051796-500MG 051796-2.5G
101897-076EA 364.62 USD
101897-076 101897-078
2-[4-(5-Bromo-4-methyl-2-pyridinyl)-1-piperazinyl]-1-ethanol
2-[4-(5-Bromo-4-methyl-2-pyridinyl)-1-piperazinyl]-1-ethanol

Hazard: Irritant

Formula: C₁₂H₁₈BrN₃O
MDL Number: MFCD13561808
CAS Number: 1220028-04-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR