5-Bromo-2-(2-ethyl-1-piperidinyl)-4-methylpyridine
Supplier: Matrix Scientific
051793-500MG
051793-2.5G
101897-064EA
364.62
USD
101897-064
101897-066
5-Bromo-2-(2-ethyl-1-piperidinyl)-4-methylpyridine
5-Bromo-2-(2-ethyl-1-piperidinyl)-4-methylpyridine
Hazard: Irritant
Formula:
C₁₃H₁₉BrN₂ |
MDL Number:
MFCD13561805 CAS Number: 1220036-19-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...