N-(5-Bromo-4-methyl-2-pyridinyl)-N-cyclohexyl-N-methylamine

Supplier: Matrix Scientific

051792-500MG 051792-2.5G
101897-060EA 364.62 USD
101897-060 101897-062
N-(5-Bromo-4-methyl-2-pyridinyl)-N-cyclohexyl-N-methylamine
N-(5-Bromo-4-methyl-2-pyridinyl)-N-cyclohexyl-N-methylamine

Hazard: Irritant

Formula: C₁₃H₁₉BrN₂
MDL Number: MFCD13561804
CAS Number: 1220029-11-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR