1-(5-Bromo-4-methyl-2-pyridinyl)-4-piperidinol

Supplier: Matrix Scientific

051790-500MG 051790-2.5G
101897-052EA 384.62 USD
101897-052 101897-054
1-(5-Bromo-4-methyl-2-pyridinyl)-4-piperidinol
1-(5-Bromo-4-methyl-2-pyridinyl)-4-piperidinol

Hazard: Irritant

Formula: C₁₁H₁₅BrN₂O
MDL Number: MFCD13561802
CAS Number: 1220019-87-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR