4-(5-Bromo-4-methyl-2-pyridinyl)-2-piperazinone
Supplier: Matrix Scientific
051789-500MG
051789-2.5G
101897-048EA
461.54
USD
101897-048
101897-050
4-(5-Bromo-4-methyl-2-pyridinyl)-2-piperazinone
4-(5-Bromo-4-methyl-2-pyridinyl)-2-piperazinone
Hazard: Irritant
Formula:
C₁₀H₁₂BrN₃O MW: 270.14 g/mol |
MDL Number:
MFCD13561801 CAS Number: 1219961-02-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...