1-(5-Bromo-4-methyl-2-pyridinyl)azepane
Supplier: Matrix Scientific
051788-500MG
051788-2.5G
101897-044EA
364.62
USD
101897-044
101897-046
1-(5-Bromo-4-methyl-2-pyridinyl)azepane
1-(5-Bromo-4-methyl-2-pyridinyl)azepane
Hazard: Irritant
Formula:
C₁₂H₁₇BrN₂ |
MDL Number:
MFCD13561800 CAS Number: 1219967-47-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...