1-Benzyl-4-(5-bromo-4-methyl-2-pyridinyl)piperazine

Supplier: Matrix Scientific

051787-500MG 051787-2.5G
101897-040EA 364.62 USD
101897-040 101897-042
1-Benzyl-4-(5-bromo-4-methyl-2-pyridinyl)piperazine
1-Benzyl-4-(5-bromo-4-methyl-2-pyridinyl)piperazine

Hazard: Irritant

Formula: C₁₇H₂₀BrN₃
MDL Number: MFCD13561799
CAS Number: 1219967-39-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR