5-Bromo-4-methyl-2-(3-methyl-1-piperidinyl)pyridine

Supplier: Matrix Scientific

051784-500MG 051784-2.5G
101897-028EA 364.62 USD
101897-028 101897-030
5-Bromo-4-methyl-2-(3-methyl-1-piperidinyl)pyridine
5-Bromo-4-methyl-2-(3-methyl-1-piperidinyl)pyridine

Hazard: Irritant

Formula: C₁₂H₁₇BrN₂
MW: 269.19 g/mol
MDL Number: MFCD13561797
CAS Number: 1220027-77-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR