5-Bromo-4-methyl-2-(2-methyl-1-piperidinyl)pyridine
Supplier: Matrix Scientific
051783-500MG
051783-2.5G
101897-024EA
364.62
USD
101897-024
101897-026
5-Bromo-4-methyl-2-(2-methyl-1-piperidinyl)pyridine
5-Bromo-4-methyl-2-(2-methyl-1-piperidinyl)pyridine
Hazard: Irritant
Formula:
C₁₂H₁₇BrN₂ MW: 269.19 g/mol |
MDL Number:
MFCD13561796 CAS Number: 1220028-97-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...