N,N-Diallyl-5-bromo-4-methyl-2-pyridinamine

Supplier: Matrix Scientific

051782-500MG 051782-2.5G
101897-020EA 364.62 USD
101897-020 101897-022
N,N-Diallyl-5-bromo-4-methyl-2-pyridinamine
N,N-Diallyl-5-bromo-4-methyl-2-pyridinamine

Hazard: Irritant

Formula: C₁₂H₁₅BrN₂
MDL Number: MFCD13561795
CAS Number: 1219960-87-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR