2-[(5-Bromo-4-methyl-2-pyridinyl)(ethyl)amino]-1-ethanol
Supplier: Matrix Scientific
051781-500MG
051781-2.5G
101897-016EA
364.62
USD
101897-016
101897-018
2-[(5-Bromo-4-methyl-2-pyridinyl)(ethyl)amino]-1-ethanol
2-[(5-Bromo-4-methyl-2-pyridinyl)(ethyl)amino]-1-ethanol
Hazard: Irritant
Formula:
C₁₀H₁₅BrN₂O |
MDL Number:
MFCD13561794 CAS Number: 1219960-92-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...