1-(5-Bromo-4-methyl-2-pyridinyl)-3-pyrrolidinol

Supplier: Matrix Scientific

051779-500MG 051779-2.5G
101897-008EA 384.62 USD
101897-008 101897-010
1-(5-Bromo-4-methyl-2-pyridinyl)-3-pyrrolidinol
1-(5-Bromo-4-methyl-2-pyridinyl)-3-pyrrolidinol

Hazard: Irritant

Formula: C₁₀H₁₃BrN₂O
MW: 257.14 g/mol
MDL Number: MFCD13561792
CAS Number: 1220028-92-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR