5-Bromo-4-methyl-2-(4-morpholino)pyridine

Supplier: Matrix Scientific

Synonyms: 4-(5-Bromo-4-methyl-2-pyridinyl)morpholine

051778-500MG 051778-2.5G
101897-004EA 184.62 USD
101897-004 101897-006
5-Bromo-4-methyl-2-(4-morpholino)pyridine
5-Bromo-4-methyl-2-(4-morpholino)pyridine

Hazard: Irritant

Formula: C₁₀H₁₃BrN₂O
MDL Number: MFCD12546513
CAS Number: 1187385-96-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR