5-Bromo-4-methyl-2-(1-piperidinyl)pyridine
Supplier: Matrix Scientific
051777-500MG
051777-2.5G
101897-000EA
364.62
USD
101897-000
101897-002
5-Bromo-4-methyl-2-(1-piperidinyl)pyridine
5-Bromo-4-methyl-2-(1-piperidinyl)pyridine
Hazard: Irritant
Formula:
C₁₁H₁₅BrN₂ |
MDL Number:
MFCD13561791 CAS Number: 1219960-78-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...