2-[(5-Bromo-4-methyl-2-pyridinyl)(methyl)amino]-1-ethanol

Supplier: Matrix Scientific

051776-500MG 051776-2.5G
101896-996EA 166.15 USD
101896-996 101896-998
2-[(5-Bromo-4-methyl-2-pyridinyl)(methyl)amino]-1-ethanol
2-[(5-Bromo-4-methyl-2-pyridinyl)(methyl)amino]-1-ethanol

Hazard: Irritant

Formula: C₉H₁₃BrN₂O
MDL Number: MFCD13561790
CAS Number: 1220027-70-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR